Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hesperetin 7-O-glucoside | 31712-49-9 | MFCD20275042 | 99.4% | 464.4 g/mol | C22H24O11 | 100MG
SDP

Catalog No. 5000238541
Encompass_Preferred

Hesperetin 7-O-glucoside is a flavanone O-glucoside used in research for its anti-inflammatory, antihypertensive, and antibacterial properties; it has reported activity against digestive enzymes and lipid-regulating targets and is provided as a research reagent.

  • Flavanone O-glucoside with molecular formula C22H24O11.
  • Molecular weight 464.4 g/mol.
  • High purity (99.4% by HPLC).
  • Reported inhibition: intestinal maltase (Ki 1.8 mM) and HMG-CoA reductase (Ki 9.8 μM).
  • Storage: store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month), protected from light.
  • Supplied for research use only; not validated for clinical use.

Catalog No. 50-002-38541 Supplier Medchemexpress LLC Supplier No. HY125130100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hesperetin 7-O-glucoside is a flavanone O-glucoside used in research for its anti-inflammatory, antihypertensive, and antibacterial properties; it has reported activity against digestive enzymes and lipid-regulating targets and is provided as a research reagent.

  • Flavanone O-glucoside with molecular formula C22H24O11.
  • Molecular weight 464.4 g/mol.
  • High purity (99.4% by HPLC).
  • Reported inhibition: intestinal maltase (Ki 1.8 mM) and HMG-CoA reductase (Ki 9.8 μM).
  • Storage: store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month), protected from light.
  • Supplied for research use only; not validated for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.