Learn More
Medchemexpress LLC Hesperetin 7-O-glucoside | 31712-49-9 | MFCD20275042 | 99.4% | 464.42 g/mol | C22H24O11 | 50MG
SDP
Catalog No. 5000238485
Encompass_Preferred
Hesperetin 7-O-glucoside is a flavanone monoglucoside derived from citrus flavonoids used in biochemical research. It demonstrates anti-inflammatory and orally active antihypertensive effects, inhibits human intestinal maltase and HMG-CoA reductase, and has reported antibacterial and gut microbiome-modulating activity. Supplied as a solid for laboratory research applications.
  • High purity suitable for research (99.4%).
  • Molecular formula C22H24O11, molecular weight 464.42 g/mol.
  • Demonstrated inhibition of intestinal maltase and HMG-CoA reductase.
  • Shows anti-inflammatory and antihypertensive activities.
  • Reported antibacterial effects and regulation of intestinal flora.
  • Available in small quantities for experimental use (1 mg-100 mg).
Catalog No. 50-002-38485 Supplier Medchemexpress LLC Supplier No. HY12513050MG
Hesperetin 7-O-glucoside is a flavanone monoglucoside derived from citrus flavonoids used in biochemical research. It demonstrates anti-inflammatory and orally active antihypertensive effects, inhibits human intestinal maltase and HMG-CoA reductase, and has reported antibacterial and gut microbiome-modulating activity. Supplied as a solid for laboratory research applications.
  • High purity suitable for research (99.4%).
  • Molecular formula C22H24O11, molecular weight 464.42 g/mol.
  • Demonstrated inhibition of intestinal maltase and HMG-CoA reductase.
  • Shows anti-inflammatory and antihypertensive activities.
  • Reported antibacterial effects and regulation of intestinal flora.
  • Available in small quantities for experimental use (1 mg-100 mg).