Learn More
TARGETMOL CHEMICALS INC Hesperetin 7-O-glucoside 5MG
SDP
Catalog No. 502879571
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Hesperetin 7-O-glucoside and prunin are direct precursors of naringin and neohesperidin, respectively, in C. aurantium. Hesperetin 7-O-glucoside shows inhibition of human HMG-CoA reductase, it also exhibits effective inhibition of the growth of Helicobacter pylori. Hesperetin 7-O-glucoside can reduce blood pressure in healthy volunteers. Purity 98.1%
Catalog No. 50-287-9571 Supplier TARGETMOL CHEMICALS INC Supplier No. TN17335MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Hesperetin 7-O-glucoside and prunin are direct precursors of naringin and neohesperidin, respectively, in C. aurantium. Hesperetin 7-O-glucoside shows inhibition of human HMG-CoA reductase, it also exhibits effective inhibition of the growth of Helicobacter pylori. Hesperetin 7-O-glucoside can reduce blood pressure in healthy volunteers. Purity 98.1%