Learn More
AdipoGen Hesperidin (100 g)
SDP
Catalog No. 502054498
Encompass_Preferred
Chemical. CAS: 520-26-3. Formula: C28H34O15. MW: 610.56. Isolated from plant source. Hesperidin is a flavanone rutinoside first isolated from citrus peels. It is metabolized by intestinal bacteria to an aglycone form, hesperetin, which is thought to be more bioavailable due to reduced polarity that allows for increased cell permeability. Both hesperidin (Hsd) and its aglycone hesperetin (Hst) have various biological properties. Studies have shown both anti-cancer and cancer chemopreventive effects, associated with their antioxidant, radical scavenging and anti-inflammatory activities. In addition, Hsd and Hst interfere at different stages of cancer. They inhibit tumor growth by targeting multiple cellular protein targets at the same time, including caspases, Bcl-2 and Bax for the induction of apoptosis, and COX-2, MMP-2 and MMP-9 for the inhibition of angiogenesis and metastasis. They also have show cardioprotective, neuroprotective and antimicrobial effects.
Catalog No. 50-205-4498 Supplier AdipoGen Supplier No. CDXH0245G100
Chemical. CAS: 520-26-3. Formula: C28H34O15. MW: 610.56. Isolated from plant source. Hesperidin is a flavanone rutinoside first isolated from citrus peels. It is metabolized by intestinal bacteria to an aglycone form, hesperetin, which is thought to be more bioavailable due to reduced polarity that allows for increased cell permeability. Both hesperidin (Hsd) and its aglycone hesperetin (Hst) have various biological properties. Studies have shown both anti-cancer and cancer chemopreventive effects, associated with their antioxidant, radical scavenging and anti-inflammatory activities. In addition, Hsd and Hst interfere at different stages of cancer. They inhibit tumor growth by targeting multiple cellular protein targets at the same time, including caspases, Bcl-2 and Bax for the induction of apoptosis, and COX-2, MMP-2 and MMP-9 for the inhibition of angiogenesis and metastasis. They also have show cardioprotective, neuroprotective and antimicrobial effects.