Learn More
Medchemexpress LLC HY-15337 10mg Medchemexpress, Hesperidin CAS:520-26-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1533710MG
Encompass_Preferred
Medchemexpress, HY-15337 10mg Hesperidin CAS:520-26-3 Hesperidin (Hesperetin 7-rutinoside), a flavanone glycoside, is isolated from citrus fruits. Hesperidin has numerous biological properties, such as decreasing inflammatory mediators and exerting significant antioxidant effects. Hesperidin also exhibits antitumor and antiallergic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1708