Learn More
Indofine Chemical Hesperidin, 98%, 5 Gm
SDP
Catalog No. 50908523
Encompass_Preferred
HESPERIDIN 5 GM, Flavanone, Hesperetin-7-O-rutinoside, C28H34O15, MW: 610.57, 98+%, 520-26-3, MFCD00075663
Catalog No. 50-908-523 Supplier Indofine Chemical Supplier No. 06017 5 GM
HESPERIDIN 5 GM, Flavanone, Hesperetin-7-O-rutinoside, C28H34O15, MW: 610.57, 98+%, 520-26-3, MFCD00075663