Learn More
Medchemexpress LLC Hesperidin | 520-26-3 | MFCD00075663 | 96.5% | 610.56 g/mol | C28H34O15 | 10 MG
SDP
Catalog No. 5000285759
Encompass_Preferred
Hesperidin (analytical standard) is a flavanone glycoside isolated from citrus fruits and supplied as a solid reference material for research and analytical applications. It is intended for method development, calibration, and quality control in chromatographic and mass spectrometric assays, and is characterized to support reliable identification and quantitation in analytical workflows.
  • Intended for analytical and research use.
  • Solid reference material for HPLC and LC-MS workflows.
  • Reported purity 96.54%, supporting accurate quantitation.
  • Includes standard chemical identifiers (CAS 520-26-3) for traceability.
  • Available in small package sizes suitable for analytical laboratories.
Catalog No. 50-002-85759 Supplier Medchemexpress LLC Supplier No. HY15337R10MG
Hesperidin (analytical standard) is a flavanone glycoside isolated from citrus fruits and supplied as a solid reference material for research and analytical applications. It is intended for method development, calibration, and quality control in chromatographic and mass spectrometric assays, and is characterized to support reliable identification and quantitation in analytical workflows.
  • Intended for analytical and research use.
  • Solid reference material for HPLC and LC-MS workflows.
  • Reported purity 96.54%, supporting accurate quantitation.
  • Includes standard chemical identifiers (CAS 520-26-3) for traceability.
  • Available in small package sizes suitable for analytical laboratories.