Learn More
Medchemexpress LLC Hesperidin | 520-26-3 | MFCD00075663 | C28H34O15 | 5 MG
SDP
Catalog No. 5000373353
Encompass_Preferred
Hesperidin (Hesperetin 7-rutinoside) is a flavanone glycoside isolated from citrus fruits. It possesses numerous biological properties, including decreasing inflammatory mediators and significant antioxidant, antitumor, and antiallergic effects.
  • Decreases inflammatory mediators
  • Significant antioxidant effects
  • Significant antitumor effects
  • Significant antiallergic effects
Catalog No. 50-003-73353 Supplier Medchemexpress LLC Supplier No. HY153375MG
Hesperidin (Hesperetin 7-rutinoside) is a flavanone glycoside isolated from citrus fruits. It possesses numerous biological properties, including decreasing inflammatory mediators and significant antioxidant, antitumor, and antiallergic effects.
  • Decreases inflammatory mediators
  • Significant antioxidant effects
  • Significant antitumor effects
  • Significant antiallergic effects