Learn More
Medchemexpress LLC Hesperidin methylchalcone | 1 g | | HY-126382-1g | 24292-52-2 | MW: 624.59
SDP
Supplier:  Medchemexpress LLC HY1263821G
Encompass_Preferred
Hesperidin methylchalcone (Hesperidin methyl chalcone) is an orally active flavonoid that has analgesic, anti-inflammatory and antioxidant properties. Hesperidin methylchalcone exhibits vasoprotective activity. Hesperidin methylchalcone inhibits oxidative stress, cytokine production and NF-κB activation. Hesperidin methylchalcone can be used for the research of gout disease.
Catalog No. NC2938719