Learn More
Medchemexpress LLC Hesperidin methylchalcone | 24292-52-2 | 98.0% | 624.59 | 10 G
SDP
Catalog No. 5000435832
Encompass_Preferred
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. It exhibits vasoprotective activity and is suitable for research into gout disease.
  • Orally active flavonoid
  • Exhibits analgesic properties
  • Acts as an anti-inflammatory agent
  • Provides antioxidant benefits
  • Shows vasoprotective activity
  • Inhibits oxidative stress
  • Reduces cytokine production
  • Suppresses NF-κB activation
  • Useful for gout disease research
Catalog No. 50-004-35832 Supplier Medchemexpress LLC Supplier No. HY12638210G
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. It exhibits vasoprotective activity and is suitable for research into gout disease.
  • Orally active flavonoid
  • Exhibits analgesic properties
  • Acts as an anti-inflammatory agent
  • Provides antioxidant benefits
  • Shows vasoprotective activity
  • Inhibits oxidative stress
  • Reduces cytokine production
  • Suppresses NF-κB activation
  • Useful for gout disease research