Learn More
Medchemexpress LLC Hesperidin methylchalcone | 24292-52-2 | 98.0% | 624.59 | 10 MM 1 ML
SDP
Catalog No. 5000435777
Encompass_Preferred
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. It exhibits vasoprotective activity and inhibits oxidative stress, cytokine production, and NF-κB activation, making it suitable for gout disease research.
  • Orally active flavonoid
  • Analgesic properties
  • Anti-inflammatory properties
  • Antioxidant properties
  • Exhibits vasoprotective activity
  • Inhibits oxidative stress
  • Inhibits cytokine production
  • Inhibits NF-κB activation
  • Suitable for gout disease research
Catalog No. 50-004-35777 Supplier Medchemexpress LLC Supplier No. HY12638210MM/1ML
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. It exhibits vasoprotective activity and inhibits oxidative stress, cytokine production, and NF-κB activation, making it suitable for gout disease research.
  • Orally active flavonoid
  • Analgesic properties
  • Anti-inflammatory properties
  • Antioxidant properties
  • Exhibits vasoprotective activity
  • Inhibits oxidative stress
  • Inhibits cytokine production
  • Inhibits NF-κB activation
  • Suitable for gout disease research