Learn More
Medchemexpress LLC Hesperidin methylchalcone | 24292-52-2 | 98.0% | 624.59 | 5 G
SDP
Catalog No. 5000438117
Encompass_Preferred
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. This compound exhibits vasoprotective activity, and it inhibits oxidative stress, cytokine production, and NF-κB activation. It can be utilized for research related to gout disease.
  • Orally active flavonoid
  • Analgesic properties
  • Anti-inflammatory properties
  • Antioxidant properties
  • Vasoprotective activity
  • Inhibits oxidative stress
  • Inhibits cytokine production
  • Inhibits NF-κB activation
  • Useful for research related to gout disease
Catalog No. 50-004-38117 Supplier Medchemexpress LLC Supplier No. HY1263825G
Hesperidin methylchalcone is an orally active flavonoid with analgesic, anti-inflammatory, and antioxidant properties. This compound exhibits vasoprotective activity, and it inhibits oxidative stress, cytokine production, and NF-κB activation. It can be utilized for research related to gout disease.
  • Orally active flavonoid
  • Analgesic properties
  • Anti-inflammatory properties
  • Antioxidant properties
  • Vasoprotective activity
  • Inhibits oxidative stress
  • Inhibits cytokine production
  • Inhibits NF-κB activation
  • Useful for research related to gout disease