Learn More
Medchemexpress LLC Hesperidin (Standard) | 520-26-3 | MFCD00006760 | C₂₈H₃₄O₁₅ | 5 MG
SDP
Catalog No. 5000373356
Encompass_Preferred
Hesperidin (Standard) is the analytical standard of Hesperidin, intended for research and analytical applications. It is a flavanone glycoside isolated from citrus fruits. Hesperidin possesses numerous biological properties, including decreasing inflammatory mediators and exhibiting significant antioxidant, antitumor, and antiallergic activities.
  • Analytical standard for hesperidin
  • Intended for research and analytical applications
  • Flavanone glycoside isolated from citrus fruits
  • Decreases inflammatory mediators
  • Exhibits significant antioxidant effects
  • Exhibits antitumor and antiallergic activities
  • Used as a reference standard in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-003-73356 Supplier Medchemexpress LLC Supplier No. HY15337R5MG
Hesperidin (Standard) is the analytical standard of Hesperidin, intended for research and analytical applications. It is a flavanone glycoside isolated from citrus fruits. Hesperidin possesses numerous biological properties, including decreasing inflammatory mediators and exhibiting significant antioxidant, antitumor, and antiallergic activities.
  • Analytical standard for hesperidin
  • Intended for research and analytical applications
  • Flavanone glycoside isolated from citrus fruits
  • Decreases inflammatory mediators
  • Exhibits significant antioxidant effects
  • Exhibits antitumor and antiallergic activities
  • Used as a reference standard in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications