Learn More
Apexbio Technology LLC HESPERITIN-50MG
SDP
Catalog No. 502240649
Encompass_Preferred
Hesperitin (CAS No 520-33-2) is a bioflavonoid primarily investigated in the fields of pharmacology and biochemistry due to its antioxidant and anti-inflammatory properties It originates from citrus fruits and acts by modulating various molecular targets and pathways including the inhibition of reactive oxygen species and suppression of inflammatory cytokines which are central to its mechanism of action In vitro Hesperitin demonstrates potency in the nanomolar to low micromolar range It is widely used in cell models and animal studies to explore its potential in mitigating oxidative stress and inflammation-related conditions Additionally it is employed in drug screening for its promising therapeutic prospects Experimental concentrations typically depend on the specific research design
This item is not returnable. View return policy
Catalog No. 50-224-0649 Supplier Apexbio Technology LLC Supplier No. N185650
Hesperitin (CAS No 520-33-2) is a bioflavonoid primarily investigated in the fields of pharmacology and biochemistry due to its antioxidant and anti-inflammatory properties It originates from citrus fruits and acts by modulating various molecular targets and pathways including the inhibition of reactive oxygen species and suppression of inflammatory cytokines which are central to its mechanism of action In vitro Hesperitin demonstrates potency in the nanomolar to low micromolar range It is widely used in cell models and animal studies to explore its potential in mitigating oxidative stress and inflammation-related conditions Additionally it is employed in drug screening for its promising therapeutic prospects Experimental concentrations typically depend on the specific research design