Learn More
Medchemexpress LLC HY-124527 10mg Medchemexpress, HET0016 CAS:339068-25-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12452710MG
Encompass_Preferred
Medchemexpress, HY-124527 10mg HET0016 CAS:339068-25-6 HET0016 is a potent and selective 20-hydroxyeicosatetraenoic acid (20-HETE) synthase inhibitor, with IC50 values of 17.7 nM, 12.1 nM and 20.6 nM for recombinant CYP4A1-, CYP4A2- and CYP4A3-catalyzed 20-HETE synthesis, respectively. HET0016 also is a selective CYP450 inhibitor, which has been shown to inhibit angiogenesis and tumor growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9666