Learn More
ATCC Het-1A; Esophageal Epithelium; Human (Homo sapiens)
Catalog No. 502385110
Encompass
Het-1A is an epithelial cell line that was isolated in 1986 from the esophagus of a normal Black, 29-year-old, male donor. This cell line was deposited by CC Harris.
Catalog No. 50-238-5110 Supplier ATCC Supplier No. CRL2692
Het-1A is an epithelial cell line that was isolated in 1986 from the esophagus of a normal Black, 29-year-old, male donor. This cell line was deposited by CC Harris.