Learn More
OMEGA BIOLOGICALS HeteroBlock, 5mg
Catalog No. NC9679584
Encompass
HeteroBlock (TM) can eliminate false positive results caused by the presence of heterophilic antibodies in a two-site sandwich immunoassay. This liquid reagent contains active agents & specific blockers yielding a stable assay signal with the use of microgram qty's.
Catalog No. NC9679584 Supplier OMEGA BIOLOGICALS Supplier No. 50011001
HeteroBlock (TM) can eliminate false positive results caused by the presence of heterophilic antibodies in a two-site sandwich immunoassay. This liquid reagent contains active agents & specific blockers yielding a stable assay signal with the use of microgram qty's.