Learn More
TARGETMOL CHEMICALS INC Heteroclitin D 5MG
SDP
Catalog No. 502878887
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1. Heteroclitin D can inhibit L-type calcium channels. 2. Heteroclitin D might be a valuable antitumor promoter or chemopreventor. Purity 99.91%
Catalog No. 50-287-8887 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S19165MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1. Heteroclitin D can inhibit L-type calcium channels. 2. Heteroclitin D might be a valuable antitumor promoter or chemopreventor. Purity 99.91%