Learn More
SCANTIBODIES LABORATORY INC HETEROPHILIC BLOCKING TUBE (3IX762) 50PK
Catalog No. NC1035777
Encompass
The 3IX762 HBT (Heterophilic Blocking Tube) from Scantibodies Laboratory, Inc. contains a unique blocking reagent composed of specific binders which inactivate heterophilic antibodies. Once the specific binders have bound to the heterophilic antibodies, the antibodies are no longer able to cause immunoassay interference. The reagent is in the form of a lyophilized pellet at the bottom of the tube and each tube contains enough reagents to inactivate the heterophilic antibodies in 500 μl of sample material. The HBT also allows for the rapid and simple elimination of false positive heterophilic interference in plasma or serum for immunoassays (i.e., FSH, LH, Prolactin, TSH, Ferritin, CEA, AFP, hCG, HBsAg, CK-MB, CA 125, CA 19-9, NSE, etc.).
Catalog No. NC1035777 Supplier SCANTIBODIES LABORATORY INC Supplier No. 31X762
The 3IX762 HBT (Heterophilic Blocking Tube) from Scantibodies Laboratory, Inc. contains a unique blocking reagent composed of specific binders which inactivate heterophilic antibodies. Once the specific binders have bound to the heterophilic antibodies, the antibodies are no longer able to cause immunoassay interference. The reagent is in the form of a lyophilized pellet at the bottom of the tube and each tube contains enough reagents to inactivate the heterophilic antibodies in 500 μl of sample material. The HBT also allows for the rapid and simple elimination of false positive heterophilic interference in plasma or serum for immunoassays (i.e., FSH, LH, Prolactin, TSH, Ferritin, CEA, AFP, hCG, HBsAg, CK-MB, CA 125, CA 19-9, NSE, etc.).