Learn More
SCANTIBODIES LABORATORY INC Heterophilic Blocking Tube
Catalog No. NC2737275
Encompass
This item must be ordered in multiples of 50 per supplier requirements
Tube, Blocking; Heterophilic; Unique blocking reagent composed of specific binders which inactivate heterophilic antibodies; Allows rapid and simple elimination of false positive heterophilic interference in plasma for sandwich immunoassays.
Catalog No. NC2737275 Supplier SCANTIBODIES LABORATORY INC Supplier No. 3IX762EACH
This item must be ordered in multiples of 50 per supplier requirements
Tube, Blocking; Heterophilic; Unique blocking reagent composed of specific binders which inactivate heterophilic antibodies; Allows rapid and simple elimination of false positive heterophilic interference in plasma for sandwich immunoassays.