Learn More
ERA WATERS CORP HETEROTROPHIC PLATE COUNT
SDP
Catalog No. NC9799042
Encompass
Each sample is one lyophilized standard containing a Heterotrophic bacteria present in the range 5-500 CFU/mL or MPN/mL. Use with the Standard Methods 9215B - Pour Plate Method, and Most Probable Number (MPN) Method (simplate).
Catalog No. NC9799042 Supplier ERA WATERS CORP Supplier No. 079
Each sample is one lyophilized standard containing a Heterotrophic bacteria present in the range 5-500 CFU/mL or MPN/mL. Use with the Standard Methods 9215B - Pour Plate Method, and Most Probable Number (MPN) Method (simplate).