Learn More
Electron Microscopy Sciences CytoSep™ Hettich Cyto-System One Funnel Tubing Chamber, 8 ml
SDP
Catalog No. 501900471
Encompass_Preferred
The CytoSep Funnel Chambers for the Hettich Cyto-System optimize lab throughput with multiple funnel options in 2, 3 or 4 funnels per slide. These funnels are perfect for every application such as CSF, viral infections, and bronchial secretions. For large volume samples such as urine.
Size: 8 ml
Catalog No. 50-190-0471 Supplier Electron Microscopy Sciences Supplier No. 6275633
The CytoSep Funnel Chambers for the Hettich Cyto-System optimize lab throughput with multiple funnel options in 2, 3 or 4 funnels per slide. These funnels are perfect for every application such as CSF, viral infections, and bronchial secretions. For large volume samples such as urine.
Size: 8 ml