Learn More
Hettich Centrifuges Rotina 380R Benchtop Centrifuge Only, 4 x 290 mL Capacity, –20/40°C Temperature Range, 115V, 50 Hz, 18" W x 16.5" H x 29.5" D
SDP
Catalog No. NC0703144 Shop All Hettich Centrifuges Products
Encompass_Preferred
THIS ITEM IS FOR THE CENTRIFUGE ONLY. THIS DOES NOT COME WITH ANY ACCESSORIES (ROTORS, BUCKETS, INSERTS, ETC.) Centrifuge up to 4 x 290 mL bottles at a time -Rugged metal housing and stainless steel centrifugation chamber -98-program memory storage with input/recall program number. The Hettich Rotina 380/380R Benchtop Centrifuges offer both a relatively small footprint and a high throughput of larger-size samples. Select speed, RCF, rotor radius, running time, acceleration, braking ramp and temperature, all through a quick-entry foil keypad. Units include storage space for up to 98 programs. Nine acceleration and braking ramps, cut-off speed or unbraked run-down can also be preselected. Units feature a sturdy metal housing and stainless steel centrifuging chamber with viewing port in the lid. Power lid lock and emergency lid-lock release ensure safe operation.
Catalog No. NC0703144 Supplier Hettich Centrifuges Supplier No. 170601
Add to Cart
Add to Cart
Request a Quote
THIS ITEM IS FOR THE CENTRIFUGE ONLY. THIS DOES NOT COME WITH ANY ACCESSORIES (ROTORS, BUCKETS, INSERTS, ETC.) Centrifuge up to 4 x 290 mL bottles at a time -Rugged metal housing and stainless steel centrifugation chamber -98-program memory storage with input/recall program number. The Hettich Rotina 380/380R Benchtop Centrifuges offer both a relatively small footprint and a high throughput of larger-size samples. Select speed, RCF, rotor radius, running time, acceleration, braking ramp and temperature, all through a quick-entry foil keypad. Units include storage space for up to 98 programs. Nine acceleration and braking ramps, cut-off speed or unbraked run-down can also be preselected. Units feature a sturdy metal housing and stainless steel centrifuging chamber with viewing port in the lid. Power lid lock and emergency lid-lock release ensure safe operation.