Learn More
Sigma Aldrich Fine Chemicals Biosciences HettichR MIKRO 200/200R ce1EA
Catalog No. 502908882
Encompass
.nbspStandard micro centrifuge.nbspAvailable in non-refrigerated (200) and refrigerated (200R) versions.nbspCapacity 24 x 1.5/2.0 mL.nbspMax RPM/RCF 14000/18626 (fixed angle).nbspChoice of 24 or 30 place rotor or PCR strip rotor or Cryo tube rotor.nbspBio-containment autoclavable phenol resistant lids available.nbspFast cool function down to 4C in 10-15 minutes.nbspCE Compliant
Catalog No. 50-290-8882 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6521211EA
Add to Cart
Add to Cart
Request a Quote
.nbspStandard micro centrifuge.nbspAvailable in non-refrigerated (200) and refrigerated (200R) versions.nbspCapacity 24 x 1.5/2.0 mL.nbspMax RPM/RCF 14000/18626 (fixed angle).nbspChoice of 24 or 30 place rotor or PCR strip rotor or Cryo tube rotor.nbspBio-containment autoclavable phenol resistant lids available.nbspFast cool function down to 4C in 10-15 minutes.nbspCE Compliant