Learn More
Sigma Aldrich Fine Chemicals Biosciences HettichR MIKRO 220/220R ce1EA
Catalog No. 502908545
Encompass
.nbspAvailable in non-refrigerated (220) and refrigerated (220R) versions.nbspAccomodates 0.2-2.0 mL microliter tubes (The refrigerated MIKRO 220R can also accomodate tubes up to 50 mL).nbspExtremely short run-up and run-down times even with rotors attaining higher speeds.nbspCentrifuge chamber of stainless steel.nbspPowered lid-locking with viewing port in lid.nbspQuick entry foil keypad.nbspLid locking and holding during rotor run.nbspLid dropping protection.nbspEmergency lid-lock release.nbspMotor-overheating protection.nbspChamber overheating protection (MIKRO 220R only).nbspImbalance switch-off.nbspAutomatic rotor recognition.nbspEasily exchangeable rotors.nbsp10 programmable memoriesRotors and other centrifuge accessories are sold separately.
Catalog No. 50-290-8545 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7240761EA
Add to Cart
Add to Cart
Request a Quote
.nbspAvailable in non-refrigerated (220) and refrigerated (220R) versions.nbspAccomodates 0.2-2.0 mL microliter tubes (The refrigerated MIKRO 220R can also accomodate tubes up to 50 mL).nbspExtremely short run-up and run-down times even with rotors attaining higher speeds.nbspCentrifuge chamber of stainless steel.nbspPowered lid-locking with viewing port in lid.nbspQuick entry foil keypad.nbspLid locking and holding during rotor run.nbspLid dropping protection.nbspEmergency lid-lock release.nbspMotor-overheating protection.nbspChamber overheating protection (MIKRO 220R only).nbspImbalance switch-off.nbspAutomatic rotor recognition.nbspEasily exchangeable rotors.nbsp10 programmable memoriesRotors and other centrifuge accessories are sold separately.