Learn More
Sigma Aldrich Fine Chemicals Biosciences HettichR ROTOFIX 32A centr1EA
Catalog No. 502908571
Encompass
.nbspEconomy universal benchtop centrifuge.nbspNon-refrigerated.nbspCapacity 4 x 100 mL.nbspMax RPM/RCF 6000/4186 (swing-out).nbspDimensions (HxWxD) 257 mm (10.0 in.) x 366 mm (14.5 in.) x 430 mm (17.0 in.).nbspRugged versatile and indispensable for routine laboratory tasks.nbspWide variety of accessories.nbspExchangeable rotors available support round bottom tubes up to 100 mL conical tubes up to 50 mL cytology up to 8 slides carriers.nbspWeight 23 kg (50.5 lbs).nbspCE Compliant
Catalog No. 50-290-8571 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6014461EA
.nbspEconomy universal benchtop centrifuge.nbspNon-refrigerated.nbspCapacity 4 x 100 mL.nbspMax RPM/RCF 6000/4186 (swing-out).nbspDimensions (HxWxD) 257 mm (10.0 in.) x 366 mm (14.5 in.) x 430 mm (17.0 in.).nbspRugged versatile and indispensable for routine laboratory tasks.nbspWide variety of accessories.nbspExchangeable rotors available support round bottom tubes up to 100 mL conical tubes up to 50 mL cytology up to 8 slides carriers.nbspWeight 23 kg (50.5 lbs).nbspCE Compliant