Learn More
Sigma Aldrich Fine Chemicals Biosciences HettichR Universal 320/3201EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7228711EA
Encompass
Standard capacity universal benchtop centrifuge.nbspAvailable in non-refrigerated (320) and refrigerated (320R) versions.nbspCapacity 4 x 200 mL (Swing-out)/6x94 mL.nbspMax RPM/RCF 15000/21382 (fixed angle).nbspTemperature Range (320R) -20C to 4C.nbspNoise dampening technology.nbspPush button open and close lid with drop protection.nbsp10 user defined settings.nbspCE CompliantSpeciality applications available.nbspCytology up to 8 slide carriers.nbspMicrotiter and deep well platesNote The UNIVERSAL 320 centrifuge offers excellent performance and a comprehensive range of accessories for virtually any clinical or research application. In addition since the UNIVERSAL 320/320R centrifuge can be used for both standard and specialized applications they are not only versatile but cost-saving as well as space saving with respect to laboratory space.Rotors and other centrifuge accessories are sold separately.
Catalog No. 50-290-8781
Only null left
Add to Cart
Request a Quote