Learn More
Creative Biomart Recombinant HEV Antigen
Supplier:  Creative Biomart HEV097
Encompass_Preferred
The E. coli derived HEV protein is fused with beta-galactosidase at the N-Terminus and contains the HEV immunodominant regions from ORF2 403-461 a.a.
Catalog No. 50-208-4475