Learn More
Creative Biomart Recombinant HEV Antigen, GST-tagged
Supplier:  Creative Biomart HEV098
Encompass_Preferred
The E. coli derived HEV protein contains the HEV immunodominant regions from ORF2 452-617 a.a. having a Mw. of 44.3 kDa. HEV ORF2 protein is fused to a 26kDa GST tag and purified by standard chromatog
Catalog No. 50-208-4476