Learn More
Creative Biomart Recombinant HEV Antigen, b-galactosidase-tagged
Supplier:  Creative Biomart HEV100
Encompass_Preferred
The E. coli derived HEV protein is fused with beta-galactosidase at the N-Terminus and contains the HEV immunodominant ORF3 92-123 a.a.
Catalog No. 50-208-4478
 Restricted Item