Learn More
Enterprise Technology Solutions Hewlett-Packard Sheet-Feed Scanner, 75 PPM/150 IPM, 600 dpi, WE/BK
SDP
Catalog No. 5000740597
Encompass_Preferred
Give your office high-volume scanning performance with this sheet-feed scanner. Capture entire documents with one-pass duplex scanning up to 150 ipm black-and-white and color. HP EveryPage allows you to capture a wide range of documents, including stacks of mixed media sizes and types. Web Jetadmin remote discovery and monitoring make it easy to identify new devices and gain insight into scanning. Scan images directly into applications with included and full-featured TWAIN and ISIS. Easily transfer scans into editable text, encrypted PDF files and more using the built-in OCR. Create one-button scan settings for recurring tasks, and make selections using the LCD control panel. Scan directly to mobile devices with the HP JetAdvantage Capture App, and then edit and save files. Load up to 80 pages into the automatic document feeder for fast, unattended scanning. Its slim construction allows convenient use in areas with limited space.
Catalog No. 50-007-40597 Supplier Enterprise Technology Solutions Supplier No. HEWL2757A
Give your office high-volume scanning performance with this sheet-feed scanner. Capture entire documents with one-pass duplex scanning up to 150 ipm black-and-white and color. HP EveryPage allows you to capture a wide range of documents, including stacks of mixed media sizes and types. Web Jetadmin remote discovery and monitoring make it easy to identify new devices and gain insight into scanning. Scan images directly into applications with included and full-featured TWAIN and ISIS. Easily transfer scans into editable text, encrypted PDF files and more using the built-in OCR. Create one-button scan settings for recurring tasks, and make selections using the LCD control panel. Scan directly to mobile devices with the HP JetAdvantage Capture App, and then edit and save files. Load up to 80 pages into the automatic document feeder for fast, unattended scanning. Its slim construction allows convenient use in areas with limited space.