Learn More
Medchemexpress LLC HY-131904A 5mg , Hex CAS:2004714-32-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131904A5MG
Encompass_Preferred
HY-131904A 5mg Hex CAS: 2004714-32-1 Hex is an enolase inhibitor, with Ki values of 74.4 nM and 269.4 nM for ENO2 and ENO1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0717