Learn More
MSC Proto Hand Hex Bit Socket, 3/8, 5/32, 1 ea
Catalog No. 501920477
Encompass
Proto hand hex bit sockets are designed for use with a 3/8 inch drive and feature a 5/32 inch hex size. These sockets are suitable for various fastening applications, providing a secure grip for hex fasteners.
  • Hex bit socket type
  • Drive size: 3/8 in
  • Hex size: 5/32 in
  • Bit length: 1-5/8 in
  • Not insulated
  • Not tether capable
  • Material: Chrome alloy steel
  • Finish: Black oxide (stem) and polished chrome
  • Overall length: 6-29/32 in
  • Not non-sparking
  • Country of origin: United States
Catalog No. 50-192-0477 Supplier MSC Supplier No. 39937610
Proto hand hex bit sockets are designed for use with a 3/8 inch drive and feature a 5/32 inch hex size. These sockets are suitable for various fastening applications, providing a secure grip for hex fasteners.
  • Hex bit socket type
  • Drive size: 3/8 in
  • Hex size: 5/32 in
  • Bit length: 1-5/8 in
  • Not insulated
  • Not tether capable
  • Material: Chrome alloy steel
  • Finish: Black oxide (stem) and polished chrome
  • Overall length: 6-29/32 in
  • Not non-sparking
  • Country of origin: United States