Learn More
Grainger Hex Bushing Forged Steel 1 1/2 x 1 in
Catalog No. 502752654
Encompass_Preferred
The Hex Bushing Forged Steel by RBC Bearings is made from forged steel with a black oxide finish. It has a 1" hex size, 1-1/4" OD, 13/16" ID, and 1/4" thickness. The bushing has a Rockwell hardness of C38-45 and operates at temperatures up to 400 degrees Fahrenheit. It is designed for use as a bushing in heavy machinery and equipment to provide wear resistance. The forged steel construction makes this hex bushing durable and impact resistant.
Catalog No. 50-275-2654 Supplier Grainger Supplier No. 14J769
The Hex Bushing Forged Steel by RBC Bearings is made from forged steel with a black oxide finish. It has a 1" hex size, 1-1/4" OD, 13/16" ID, and 1/4" thickness. The bushing has a Rockwell hardness of C38-45 and operates at temperatures up to 400 degrees Fahrenheit. It is designed for use as a bushing in heavy machinery and equipment to provide wear resistance. The forged steel construction makes this hex bushing durable and impact resistant.