Learn More
Grainger Hex Bushing Forged Steel 1/2 x 1/4 in
Catalog No. 502749559
Encompass_Preferred
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3 inches long x 1-3/8 inches wide x 1-1/4 inches high. It weighs approximately 1 pound and has a black oxide finish. The Anvil Hex Bushing provides an easy way to convert threaded pipe to accept rods or shafts.
Catalog No. 50-274-9559 Supplier Grainger Supplier No. 14J759
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3 inches long x 1-3/8 inches wide x 1-1/4 inches high. It weighs approximately 1 pound and has a black oxide finish. The Anvil Hex Bushing provides an easy way to convert threaded pipe to accept rods or shafts.