Learn More
Grainger Hex Bushing Forged Steel 1/2 x 3/8 in
Catalog No. 502748810
Encompass_Preferred
The Hex Bushing Forged Steel from Grainger is a forged steel bushing with a hexagonal bore. It has a 1-1/4 inch hex size, 1-1/2 inch OD, 1-1/8 inch length, and is made from 1045 carbon steel. This hex bushing can be used with a hex head cap screw as an anti-rotation device or a guide. It is rated at 60,000 psi tensile strength. The bushing has a black oxide finish to resist corrosion. It meets ASTM A108 standards.
Catalog No. 50-274-8810 Supplier Grainger Supplier No. 14J760
The Hex Bushing Forged Steel from Grainger is a forged steel bushing with a hexagonal bore. It has a 1-1/4 inch hex size, 1-1/2 inch OD, 1-1/8 inch length, and is made from 1045 carbon steel. This hex bushing can be used with a hex head cap screw as an anti-rotation device or a guide. It is rated at 60,000 psi tensile strength. The bushing has a black oxide finish to resist corrosion. It meets ASTM A108 standards.