Learn More
Grainger Hex Bushing Forged Steel 1 x 1/4 in
Catalog No. 502749560
Encompass_Preferred
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90. It is designed for use as a bushing in hexagonal holes and has a pressure rating of 2,000 PSI at 100 degrees Fahrenheit. The bushing measures 1.26 inches across the flats with an inside diameter of 1.25 inches and an outside diameter of 1.5 inches. It weighs approximately 0.2 pounds.
Catalog No. 50-274-9560 Supplier Grainger Supplier No. 14J764
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90. It is designed for use as a bushing in hexagonal holes and has a pressure rating of 2,000 PSI at 100 degrees Fahrenheit. The bushing measures 1.26 inches across the flats with an inside diameter of 1.25 inches and an outside diameter of 1.5 inches. It weighs approximately 0.2 pounds.