Learn More
Grainger Hex Bushing Forged Steel 1 x 1/2 in
Catalog No. 502748812
Encompass_Preferred
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3-3/4 inch length x 2 inch height x 1-3/4 inch width. It weighs approximately 2 pounds. This heavy duty hex bushing is designed for use in pipe connections and mechanical applications requiring a hex shaped bushing.
Catalog No. 50-274-8812 Supplier Grainger Supplier No. 14J766
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3-3/4 inch length x 2 inch height x 1-3/4 inch width. It weighs approximately 2 pounds. This heavy duty hex bushing is designed for use in pipe connections and mechanical applications requiring a hex shaped bushing.