Learn More
Grainger Hex Bushing Forged Steel 1 x 3/4 in
Catalog No. 502751619
Encompass_Preferred
The Hex Bushing Forged Steel is a hexagonal bushing made of forged steel. It has a 1-1/4 inch hex size, 1-1/4 inch ID, 1-1/2 inch OD, and is 1 inch long. The bushing is made in the USA and is zinc plated for corrosion resistance. It has a Rockwell hardness of B80-90 for durability. The hex bushing can be used for shaft alignment and reducing friction and wear in machinery. Its forged steel construction makes it strong, and the hex shape allows the use of wrenches for installation and removal.
Catalog No. 50-275-1619 Supplier Grainger Supplier No. 14J767
The Hex Bushing Forged Steel is a hexagonal bushing made of forged steel. It has a 1-1/4 inch hex size, 1-1/4 inch ID, 1-1/2 inch OD, and is 1 inch long. The bushing is made in the USA and is zinc plated for corrosion resistance. It has a Rockwell hardness of B80-90 for durability. The hex bushing can be used for shaft alignment and reducing friction and wear in machinery. Its forged steel construction makes it strong, and the hex shape allows the use of wrenches for installation and removal.