Learn More
Grainger Hex Bushing Forged Steel 2 x 1/2 in
Catalog No. 502750953
Encompass_Preferred
The Anvil Hex Bushing is a forged steel hex bushing with a 1-1/4" nominal pipe size (NPS), 1-1/4" actual inside diameter, and 1-7/16" actual outside diameter. It has a pressure rating of 2,000 psi at 100°F. The bushing is made from A105 forged carbon steel and has beveled ends. It can be used with schedule 40 or 80 steel pipe and is designed for general purpose applications such as connecting pipes. The Anvil hex bushing is made in the USA and exceeds ASTM A105 requirements.
Catalog No. 50-275-0953 Supplier Grainger Supplier No. 14J772
The Anvil Hex Bushing is a forged steel hex bushing with a 1-1/4" nominal pipe size (NPS), 1-1/4" actual inside diameter, and 1-7/16" actual outside diameter. It has a pressure rating of 2,000 psi at 100°F. The bushing is made from A105 forged carbon steel and has beveled ends. It can be used with schedule 40 or 80 steel pipe and is designed for general purpose applications such as connecting pipes. The Anvil hex bushing is made in the USA and exceeds ASTM A105 requirements.