Learn More
Grainger Hex Bushing Forged Steel 2 x 1 1/2 in
Catalog No. 502750938
Encompass_Preferred
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3 inches long x 1-3/4 inches wide and it weighs 0.75 lbs. This hex bushing provides a heavy wall, precision bearing surface.
Catalog No. 50-275-0938 Supplier Grainger Supplier No. 14J776
The Anvil Hex Bushing is a forged steel bushing with a hexagonal shape. It has a 1-1/4 inch nominal pipe size and is made from 1045 carbon steel. The bushing has a Rockwell hardness of B80-90 and meets ASTM A108 standards. It can withstand temperatures up to 1000°F. The bushing's dimensions are 3 inches long x 1-3/4 inches wide and it weighs 0.75 lbs. This hex bushing provides a heavy wall, precision bearing surface.