Learn More
Grainger Hex Bushing Forged Steel 2 x 1/4 in
Catalog No. 502751620
Encompass_Preferred
The Anvil Hex Bushing is made of forged steel with a black oxide finish. It has a hex head with a 1-1/4 inch width across flats and a 1-1/4 inch-12 UN thread. The overall length is 2 inches. This hex bushing is rated at 60,000 psi tensile strength and can withstand temperatures up to 1000°F. It is designed for use as a bushing in machinery, equipment and other threaded applications requiring high strength and durability. The black oxide finish provides mild corrosion resistance.
Catalog No. 50-275-1620 Supplier Grainger Supplier No. 14J771
The Anvil Hex Bushing is made of forged steel with a black oxide finish. It has a hex head with a 1-1/4 inch width across flats and a 1-1/4 inch-12 UN thread. The overall length is 2 inches. This hex bushing is rated at 60,000 psi tensile strength and can withstand temperatures up to 1000°F. It is designed for use as a bushing in machinery, equipment and other threaded applications requiring high strength and durability. The black oxide finish provides mild corrosion resistance.