Learn More
Grainger Hex Bushing Forged Steel 2 x 3/4 in
Catalog No. 502750954
Encompass_Preferred
The Anvil Hex Bushing is a forged steel bushing with a hex head. It has a 1-1/4 inch hex size and is made from 1045 carbon steel for strength and durability. The bushing has a Rockwell hardness of C35-45 and is zinc plated for corrosion resistance. It has a temperature rating up to 450°F. The bushing has coarse, rolled threads and meets ASME B18.2.1 specifications. It is manufactured in the USA by Anvil International.
Catalog No. 50-275-0954 Supplier Grainger Supplier No. 14J773
The Anvil Hex Bushing is a forged steel bushing with a hex head. It has a 1-1/4 inch hex size and is made from 1045 carbon steel for strength and durability. The bushing has a Rockwell hardness of C35-45 and is zinc plated for corrosion resistance. It has a temperature rating up to 450°F. The bushing has coarse, rolled threads and meets ASME B18.2.1 specifications. It is manufactured in the USA by Anvil International.