Learn More
Grainger Hex Bushing Forged Steel 3/4 x 1/2 in
Catalog No. 502748811
Encompass_Preferred
The Anvil Hex Bushing Forged Steel 14J763 is a forged steel hex bushing with a 1-1/4 inch nominal pipe size (NPS). It has a 1-1/4 inch female NPT threaded connection on one end and a 1-1/4 inch male NPT threaded connection on the other end. The bushing allows you to change pipe thread sizes. It is made from forged 1035 carbon steel for durability. The maximum pressure rating is up to 5,000 psi and temperature range is -20 to 300 degrees Fahrenheit.
Catalog No. 50-274-8811 Supplier Grainger Supplier No. 14J763
The Anvil Hex Bushing Forged Steel 14J763 is a forged steel hex bushing with a 1-1/4 inch nominal pipe size (NPS). It has a 1-1/4 inch female NPT threaded connection on one end and a 1-1/4 inch male NPT threaded connection on the other end. The bushing allows you to change pipe thread sizes. It is made from forged 1035 carbon steel for durability. The maximum pressure rating is up to 5,000 psi and temperature range is -20 to 300 degrees Fahrenheit.