Learn More
Grainger Hex Coupling, Brass
Catalog No. NC2574896
Encompass_Preferred
Coupling Brass 13Y875 is a hex coupling made of brass with a zinc plating. It has a 1/4 inch NPT female pipe thread on both ends and is designed for use in instrumentation, hydraulic, and pneumatic applications. This hex coupling has a maximum pressure rating of 3000 psi at 70°F and is RoHS compliant. The dimensions are 15/16 inch hex by 1-1/4 inch long with 1/4 inch NPT threads. This brass hex coupling provides a means of connecting two male threaded pipes together in a straight configuration.
Catalog No. NC2574896 Supplier Grainger Supplier No. 13Y875
Coupling Brass 13Y875 is a hex coupling made of brass with a zinc plating. It has a 1/4 inch NPT female pipe thread on both ends and is designed for use in instrumentation, hydraulic, and pneumatic applications. This hex coupling has a maximum pressure rating of 3000 psi at 70°F and is RoHS compliant. The dimensions are 15/16 inch hex by 1-1/4 inch long with 1/4 inch NPT threads. This brass hex coupling provides a means of connecting two male threaded pipes together in a straight configuration.