Learn More
Medchemexpress LLC Hex-[Cys(tMeBn(H-AET))-Pro-Pro-Thr-Gln-Phe-Cys]-OH | 99.3% | 1198.40 | 1 MG
SDP
Catalog No. 5000398288
Encompass_Preferred
3BP-4089 TFA is a highly potent fibroblast activation protein (FAP)-targeting peptide for theranostics. It is often coupled with radionuclides for tumor diagnosis and research.
  • Highly potent FAP-targeting peptide
  • Used for theranostics
  • Can be coupled with radionuclides
  • Suitable for tumor diagnosis and research
Catalog No. 50-003-98288 Supplier Medchemexpress LLC Supplier No. HYP3441A1MG
3BP-4089 TFA is a highly potent fibroblast activation protein (FAP)-targeting peptide for theranostics. It is often coupled with radionuclides for tumor diagnosis and research.
  • Highly potent FAP-targeting peptide
  • Used for theranostics
  • Can be coupled with radionuclides
  • Suitable for tumor diagnosis and research