Learn More
Electron Microscopy Sciences Veco Hexagonal with Handle 150 mesh Copper Grid
SDP
Catalog No. 5026425
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm
Catalog No. 50-264-25 Supplier Electron Microscopy Sciences Supplier No. HD150HCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm