Learn More
Electron Microscopy Sciences Veco Hexagonal with Handle 200 mesh Copper Grid
SDP
Catalog No. 5026433
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
Catalog No. 50-264-33 Supplier Electron Microscopy Sciences Supplier No. HD200HCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm