Learn More
Electron Microscopy Sciences Veco Hexagonal with Handle 200 mesh Nickel Grid
SDP
Catalog No. 5026432
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Nickel
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
Catalog No. 50-264-32 Supplier Electron Microscopy Sciences Supplier No. HD200HNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Nickel
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm