Learn More
Electron Microscopy Sciences Veco Hexagonal with Handle 300 mesh Copper Grid
SDP
Catalog No. 5026434
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 83 µm
Hole: 45 µm
Bar: 38 µm
Catalog No. 50-264-34 Supplier Electron Microscopy Sciences Supplier No. HD300HCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Copper
Pitch: 83 µm
Hole: 45 µm
Bar: 38 µm